4-[2-Nitro-4-(trifluoromethyl)phenoxy]benzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: 4-(4-Boronophenoxy)-3-nitrobenzotrifluoride, 4-Boronophenyl 2-nitro-4-(trifluoromethyl)phenyl ether, (4-(2-Nitro-4-(trifluoromethyl)phenoxy)phenyl)boronic acid

PC8330-1G
APOSPC8330-1GEA 125 EUR
APOSPC8330-1G
4-[2-Nitro-4-(trifluoromethyl)phenoxy]benzeneboronic acid
4-[2-Nitro-4-(trifluoromethyl)phenoxy]benzeneboronic acid
Formel: C₁₃H₉BF₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD09800896
CAS-Nummer: 957062-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR