3-Cyan-2-fluorphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Cyano-2-fluorophenyl)boronic acid
PC8333-1G
APOSPC8333-1GEA
34.8
EUR
APOSPC8333-1G
3-Cyan-2-fluorphenylboronsäure
3-Cyan-2-fluorphenylboronsäure
Formel:
C₇H₅BFNO₂ Molecular Weight: 164,9295 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07374984 CAS-Nummer: 957121-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...