Methyl-5-(4-fluorphenyl)-4-oxazolcarboxylat

Lieferant: Apollo Scientific

PC8338-1G
APOSPC8338-1GEA 314 EUR
APOSPC8338-1G
Methyl-5-(4-fluorphenyl)-4-oxazolcarboxylat
Methyl-5-(4-fluorphenyl)-4-oxazolcarboxylat
Formel: C₁₁H₈ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09800994
CAS-Nummer: 89204-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR