4-Brom-2-fluorphenylessigsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Bromo-2-fluorophenyl)acetic acid
PC8343-25G
APOSPC8343-25GEA
65.4
EUR
APOSPC8343-25G
4-Brom-2-fluorphenylessigsäure
4-Brom-2-fluorphenylessigsäure
Formel:
C₈H₆BrFO₂ Molecular Weight: 233.03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09032952 CAS-Nummer: 114897-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...