2-(4-Fluorobenzylthio)ethylamine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC8344-100MG
PC8344-500MG
APOSPC8344-100MGEA
221
EUR
APOSPC8344-100MG
APOSPC8344-500MG
2-(4-Fluorobenzylthio)ethylamine 95%
2-(4-Fluorobenzylthio)ethylamine
Formel:
C₉H₁₂FNS Siedepunkt: 98 °C (0,5 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01764909 CAS-Nummer: 143627-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...