O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 97%
Lieferant: Apollo Scientific
Synonyme:
O-(1H-6-Chlorobenzotriazol-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate, TCTU
PC8348-100G
APOSPC8348-100GEA
71.2
EUR
APOSPC8348-100G
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 97%
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate
Formel:
C₁₁H₁₅BClF₄N₅O Molecular Weight: 355,53 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973270 CAS-Nummer: 330641-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...