N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate

PC8350-100G PC8350-25G PC8350-5G
APOSPC8350-100GEA 69.6 EUR
APOSPC8350-100G APOSPC8350-25G APOSPC8350-5G
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 98%
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat
Formel: C₉H₁₆BF₄N₃O₃
Molecular Weight: 301.05 g/mol
Schmelzpunkt: 195…201 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077875
CAS-Nummer: 105832-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR