N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate
PC8350-100G
PC8350-25G
PC8350-5G
APOSPC8350-100GEA
69.6
EUR
APOSPC8350-100G
APOSPC8350-25G
APOSPC8350-5G
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 98%
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat
Formel:
C₉H₁₆BF₄N₃O₃ Molecular Weight: 301.05 g/mol Schmelzpunkt: 195…201 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00077875 CAS-Nummer: 105832-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...