N-Boc-2-trifluoromethyl-D-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluoromethyl)-D-phenylalanine, N-BOC protected, (R)-2-((tert-butoxycarbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid

PC8355-1G PC8355-25G PC8355-5G
APOSPC8355-1GEA 121 EUR
APOSPC8355-1G APOSPC8355-25G APOSPC8355-5G
N-Boc-2-trifluoromethyl-D-phenylalanine
N-Boc-2-trifluoromethyl-D-phenylalanine
Formel: C₁₅H₁₈F₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01862945
CAS-Nummer: 346694-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR