Fmoc-2-(trifluoromethyl)-L-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-Phe(2-CF₃)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid, 2-(Trifluoromethyl)-L-phenylalanine, N-FMOC protected

PC8356-5G
APOSPC8356-5GEA 229 EUR
APOSPC8356-5G
Fmoc-2-(trifluoromethyl)-L-phenylalanine
Fmoc-2-(trifluoromethyl)-L-phenylalanine
Formel: C₂₅H₂₀F₃NO₄
Molecular Weight: 455.44 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863052
CAS-Nummer: 352523-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR