2-Fluor-D-phenylalanin

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-3-(2-fluorophenyl)propanoic acid, H-D-Phe(2-F)-OH

PC8358-5G
APOSPC8358-5GEA 68.3 EUR
APOSPC8358-5G
2-Fluor-D-phenylalanin
2-Fluor-D-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066447
CAS-Nummer: 97731-02-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR