Pentafluoro-L-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-3-(perfluorophenyl)propanoic acid
PC8360-1G
APOSPC8360-1GEA
132
EUR
APOSPC8360-1G
Pentafluoro-L-phenylalanine
Pentafluoro-L-phenylalanine
Formel:
C₉H₆F₅NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01860881 CAS-Nummer: 34702-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...