D-Pentafluorophenylalanine

Lieferant: Apollo Scientific

Synonyme: (2R)-2-Amino-3-(pentafluorophenyl)propanoic acid, (R)-2-Amino-3-(perfluorophenyl)propanoic acid

PC8361-1G
APOSPC8361-1GEA 136 EUR
APOSPC8361-1G
D-Pentafluorophenylalanine
D-Pentafluorophenylalanine
Formel: C₉H₆F₅NO₂
Molecular Weight: 255,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860872
CAS-Nummer: 40332-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR