2-(Trifluoromethyl)-D-phenyalanine 97+%
Lieferant: Apollo Scientific
Synonyme:
2-(Trifluoromethyl)-D-phenylalanine, H-D-Phe(2-CF₃)-OH
PC8366-1G
APOSPC8366-1GEA
87
EUR
APOSPC8366-1G
2-(Trifluoromethyl)-D-phenyalanine 97+%
2-(Trifluoromethyl)-D-phenyalanine
Formel:
C₁₀H₁₀F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01860877 CAS-Nummer: 130930-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...