5-Brom-2-fluor-4-methylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC8382-5G
APOSPC8382-5GEA
26.1
EUR
APOSPC8382-5G
5-Brom-2-fluor-4-methylpyridin
5-Brom-2-fluor-4-methylpyridin
Formel:
C₆H₅BrFN Molecular Weight: 190,01 g/mol Dichte: 1,62 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839213 CAS-Nummer: 864830-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...