2-Chloro-5-fluoro-4-methylpyridine 97+%

Lieferant: Apollo Scientific

PC8383-5G
APOSPC8383-5GEA 62.5 EUR
APOSPC8383-5G
2-Chloro-5-fluoro-4-methylpyridine 97+%
2-Chloro-5-fluoro-4-methylpyridine
Formel: C₆H₅ClFN
Molecular Weight: 145,5600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864695
CAS-Nummer: 881891-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR