2-Brom-2',4'-bis(trifluormethyl)acetophenon
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,4-Bis(trifluoromethyl)phenacyl bromide
PC8402-1G
PC8402-5G
APOSPC8402-1GEA
292
EUR
APOSPC8402-1G
APOSPC8402-5G
2-Brom-2',4'-bis(trifluormethyl)acetophenon
2-Brom-2',4'-bis(trifluormethyl)acetophenon
Formel:
C₁₀H₅BrF₆O Schmelzpunkt: 50…52 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094502 CAS-Nummer: 435271-21-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...