2-Brom-4'-fluor-3'-(trifluormethyl)acetophenon 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Fluoro-3-(trifluoromethyl)phenacyl bromide, 2-Bromo-1-(4-fluoro-3-(trifluoromethyl)phenyl)ethanone
PC8410-1G
PC8410-25G
PC8410-5G
APOSPC8410-1GEA
21.9
EUR
APOSPC8410-1G
APOSPC8410-25G
APOSPC8410-5G
2-Brom-4'-fluor-3'-(trifluormethyl)acetophenon 95%
2-Brom-4'-fluor-3'-(trifluormethyl)acetophenon
Formel:
C₉H₅BrF₄O Molecular Weight: 285.0330 g/mol Schmelzpunkt: 42…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094509 CAS-Nummer: 537050-14-9 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...