2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]malononitrile

Lieferant: Apollo Scientific
Achtung

PC8411-500MG
APOSPC8411-500MGEA 102 EUR
APOSPC8411-500MG
2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]malononitrile
2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]malononitrile
Formel: C₉H₃ClF₃N₃
Molecular Weight: 245,59 g/mol
Schmelzpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01315462
CAS-Nummer: 338956-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR