2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8415-1G
APOSPC8415-1GEA
66.7
EUR
APOSPC8415-1G
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 97%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline
Formel:
C₁₂H₈ClF₃N₂O Molecular Weight: 288,66 g/mol Schmelzpunkt: 95…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315051 CAS-Nummer: 98968-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...