4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-buten-1-ol 97%

Lieferant: Apollo Scientific
Achtung

PC8417-500MG
APOSPC8417-500MGEA 58 EUR
APOSPC8417-500MG
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-buten-1-ol 97%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-buten-1-ol
Formel: C₁₀H₉ClF₃NO₂
Molecular Weight: 267,64 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038247
CAS-Nummer: 338772-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR