3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}-3-iminopropionitrile 97%

Lieferant: Apollo Scientific

PC8425-500MG
APOSPC8425-500MGEA 688 EUR
APOSPC8425-500MG
3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}-3-iminopropionitrile 97%
3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}-3-iminopropionitrile
Formel: C₁₃H₁₃ClF₃N₅
Molecular Weight: 331,724 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664816
CAS-Nummer: 339103-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR