3-Amino-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]pyridine hydrochloride 97%

Lieferant: Apollo Scientific

PC8426-500MG
APOSPC8426-500MGEA 688 EUR
APOSPC8426-500MG
3-Amino-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]pyridine hydrochloride 97%
3-Amino-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]pyridine hydrochloride
Formel: C₁₅H₁₅ClF₃N₅·ClH
Molecular Weight: 394,222 g/mol
Schmelzpunkt: 200…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793058
CAS-Nummer: 317822-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR