8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-8-chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine

PC8437-1G
APOSPC8437-1GEA 66.7 EUR
APOSPC8437-1G
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridin-3-amine
Formel: C₇H₄ClF₃N₄
Molecular Weight: 236,58 g/mol
Schmelzpunkt: 273…275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00974657
CAS-Nummer: 338773-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR