2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Cyano-3-fluorophenyl)-2-(3,4-dimethoxyphenyl)acetonitrile

PC8439-500MG
APOSPC8439-500MGEA 21.9 EUR
APOSPC8439-500MG
2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile 97%
2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile
Formel: C₁₇H₁₃FN₂O₂
Molecular Weight: 296,3 g/mol
Schmelzpunkt: 160…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00231998
CAS-Nummer: 339012-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR