2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyano-N-(3-(trifluoromethyl)phenyl)acetamide, 2-Cyano-3'-(trifluoromethyl)acetanilide

PC8448-1G
APOSPC8448-1GEA 43.5 EUR
APOSPC8448-1G
2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide 97%
2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide
Formel: C₁₀H₇F₃N₂O
Molecular Weight: 228,18 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00117941
CAS-Nummer: 1960-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR