Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8453-1G
APOSPC8453-1GEA
132
EUR
APOSPC8453-1G
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate 97%
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate
Formel:
C₁₃H₁₃ClF₃NO₄ Molecular Weight: 339,7 g/mol Siedepunkt: 120…124 °C/0,5mmHg Schmelzpunkt: 43…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00231883 CAS-Nummer: 172527-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...