Diethyl 3-{[3-(trifluoromethyl)phenyl]carbamoyl}-1,2-cyclopropanedicarboxylate 97%

Lieferant: Apollo Scientific

PC8454-500MG
APOSPC8454-500MGEA 782 EUR
APOSPC8454-500MG
Diethyl 3-{[3-(trifluoromethyl)phenyl]carbamoyl}-1,2-cyclopropanedicarboxylate 97%
Diethyl 3-{[3-(trifluoromethyl)phenyl]carbamoyl}-1,2-cyclopropanedicarboxylate
Formel: C₁₇H₁₈F₃NO₅
Molecular Weight: 373,324 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01315628
CAS-Nummer: 318237-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR