3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile 97%

Lieferant: Apollo Scientific

PC8458-1G PC8458-500MG PC8458-5G
APOSPC8458-1GEA 172 EUR
APOSPC8458-1G APOSPC8458-500MG APOSPC8458-5G
3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile 97%
3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]acrylonitrile
Formel: C₁₂H₁₁F₃N₂
Schmelzpunkt: 104…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215017
CAS-Nummer: 62738-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR