2,6-Dinitro-4-(trifluormethyl)anilin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3,5-dinitro-α,α,α-trifluortoluol, 2,6-Dinitro-α,α,α-trifluor-p-toluidin

PC8461-5G
APOSPC8461-5GEA 161 EUR
APOSPC8461-5G
2,6-Dinitro-4-(trifluormethyl)anilin 97%
2,6-Dinitro-4-(trifluormethyl)anilin
Formel: C₇H₄F₃N₃O₄
Molecular Weight: 251,12 g/mol
Schmelzpunkt: 144…149 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00024260
CAS-Nummer: 445-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR