1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylic acid 97%
Lieferant: Apollo Scientific
PC8463-1G
APOSPC8463-1GEA
1450
EUR
APOSPC8463-1G
1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylic acid 97%
1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylic acid
Formel:
C₁₃H₁₂F₃N₃O₆ Molecular Weight: 363,246 g/mol Schmelzpunkt: 164…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664473 CAS-Nummer: 303144-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...