5-Brom-2,3-difluoranisol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1,2-difluoro-3-methoxybenzene
PC8472-100G
APOSPC8472-100GEA
155
EUR
APOSPC8472-100G
5-Brom-2,3-difluoranisol 97%
5-Brom-2,3-difluoranisol
Formel:
C₇H₅BrF₂O Molecular Weight: 223.02 g/mol Schmelzpunkt: 33…36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631352 CAS-Nummer: 261762-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...