4-Methoxy-3-(trifluormethyl)benzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC8475-25G
PC8475-5G
PC8475-100G
APOSPC8475-25GEA
52.2
EUR
APOSPC8475-25G
APOSPC8475-5G
APOSPC8475-100G
4-Methoxy-3-(trifluormethyl)benzoesäure
4-Methoxy-3-(trifluoromethyl)benzoic acid
Formel:
C₉H₇F₃O₃ Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01091014 CAS-Nummer: 213598-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...