Ethyl 4-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}carbonyl)-5-methylisoxazole-3-carboxylate 97%

Lieferant: Apollo Scientific

PC8477-100MG
APOSPC8477-100MGEA 288 EUR
APOSPC8477-100MG
Ethyl 4-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}carbonyl)-5-methylisoxazole-3-carboxylate 97%
Ethyl 4-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}carbonyl)-5-methylisoxazole-3-carboxylate
Formel: C₁₄H₁₁ClF₃N₃O₄
Molecular Weight: 377,703 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664738
CAS-Nummer: 338397-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR