2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide
Lieferant: Apollo Scientific
|
Achtung
|
PC8478-25G
PC8478-5G
PC8478-1G
APOSPC8478-25GEA
0
EUR
APOSPC8478-25G
APOSPC8478-5G
APOSPC8478-1G
2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide
2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide
Formel:
C₉H₇BrF₃NO Schmelzpunkt: 156…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662747 CAS-Nummer: 3823-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...