1-(4-Bromo-2-fluorophenyl)-1H-pyrrole-2,5-dione
Lieferant: Apollo Scientific
|
Achtung
|
PC8480-1G
APOSPC8480-1GEA
93.1
EUR
APOSPC8480-1G
1-(4-Bromo-2-fluorophenyl)-1H-pyrrole-2,5-dione
1-(4-Bromo-2-fluorophenyl)-1H-pyrrole-2,5-dione
Formel:
C₁₀H₅BrFNO₂ Schmelzpunkt: 97…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08277325 CAS-Nummer: 893614-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...