N-(2-Bromo-4-fluorophenyl)thiourea

Lieferant: Apollo Scientific

PC8481-1G
APOSPC8481-1GEA 270 EUR
APOSPC8481-1G
N-(2-Bromo-4-fluorophenyl)thiourea
N-(2-Bromo-4-fluorophenyl)thiourea
Formel: C₇H₆BrFN₂S
Molecular Weight: 249,11 g/mol
Schmelzpunkt: 163…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027194
CAS-Nummer: 952182-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR