2,6-Difluoro-N-hydroxybenzenecarboximidoylchloride 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC8484-1G
APOSPC8484-1GEA
348
EUR
APOSPC8484-1G
2,6-Difluoro-N-hydroxybenzenecarboximidoylchloride 95+%
2,6-Difluoro-N-hydroxybenzenecarboximidoylchloride
Formel:
C₇H₄ClF₂NO Schmelzpunkt: 104…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027155 CAS-Nummer: 118591-69-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...