N-(4-Fluoro-3-nitrophenyl)-2-(1-pyrrolidinyl)acetamide

Lieferant: Apollo Scientific

PC8485-500MG
APOSPC8485-500MGEA 69.8 EUR
APOSPC8485-500MG
N-(4-Fluoro-3-nitrophenyl)-2-(1-pyrrolidinyl)acetamide
N-(4-Fluoro-3-nitrophenyl)-2-(1-pyrrolidinyl)acetamide
Formel: C₁₂H₁₄FN₃O₃
Molecular Weight: 267,26 g/mol
Schmelzpunkt: 105…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027123
CAS-Nummer: 952182-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR