4-[(4-Fluorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde

Lieferant: Apollo Scientific

Synonyme: 4-[(4-Fluorophenyl)thio]-3-nitrobenzaldehyde

PC8488-1G
APOSPC8488-1GEA 93.1 EUR
APOSPC8488-1G
4-[(4-Fluorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde
4-[(4-Fluorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde
Formel: C₁₃H₈FNO₃S
Molecular Weight: 277,28 g/mol
Schmelzpunkt: 151…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027121
CAS-Nummer: 952182-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR