N-Hydroxy-2-(trifluoromethyl)benzenecarboximidoylchloride
Lieferant: Apollo Scientific
|
Achtung
|
PC8497-1G
APOSPC8497-1GEA
219
EUR
APOSPC8497-1G
N-Hydroxy-2-(trifluoromethyl)benzenecarboximidoylchloride
N-Hydroxy-2-(trifluoromethyl)benzenecarboximidoylchloride
Formel:
C₈H₅ClF₃NO Molecular Weight: 223.58 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027157 CAS-Nummer: 74467-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...