2-Brom-3'-fluor-4'-(trifluormethyl)acetophenon
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-4-(trifluoromethyl)phenacyl bromide
PC8501-1G
APOSPC8501-1GEA
719
EUR
APOSPC8501-1G
2-Brom-3'-fluor-4'-(trifluormethyl)acetophenon
2-Brom-3'-fluor-4'-(trifluormethyl)acetophenon
Formel:
C₉H₅BrF₄O Schmelzpunkt: 29…31 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094511 CAS-Nummer: 544429-22-3 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...