Pentafluorophenyl nicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: Pentafluorophenyl pyridine-3-carboxylate

PC8504-1G PC8504-250MG
APOSPC8504-1GEA 203 EUR
APOSPC8504-1G APOSPC8504-250MG
Pentafluorophenyl nicotinate
Pentafluorophenyl nicotinate
Formel: C₁₂H₄F₅NO₂
Schmelzpunkt: 65,5…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064938
CAS-Nummer: 848347-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR