3-(Trifluoromethylthio)phenacyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC8507-1G
PC8507-5G
APOSPC8507-1GEA
0
EUR
APOSPC8507-1G
APOSPC8507-5G
3-(Trifluoromethylthio)phenacyl bromide
3-(Trifluoromethylthio)phenacyl bromide
Formel:
C₉H₆BrF₃OS Schmelzpunkt: 32…34 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094515 CAS-Nummer: 56773-34-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...