Pentafluorophenyl 3-(pyridin-2-yloxy)benzoate 97%

Lieferant: Apollo Scientific

PC8509-1G PC8509-250MG
APOSPC8509-1GEA 220 EUR
APOSPC8509-1G APOSPC8509-250MG
Pentafluorophenyl 3-(pyridin-2-yloxy)benzoate 97%
Pentafluorophenyl 3-(pyridin-2-yloxy)benzoate
Formel: C₁₈H₈F₅NO₃
Molecular Weight: 381.253 g/mol
Schmelzpunkt: 60.5…61.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064956
CAS-Nummer: 921938-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR