2,2-Difluorobenzo[1,3]dioxole-4-boronic acid 98%

Lieferant: Apollo Scientific
Achtung

PC8515-1G
APOSPC8515-1GEA 122 EUR
APOSPC8515-1G
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid 98%
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid
Formel: C₇H₅BF₂O₄
Lagertemperatur: Kühlschrank
MDL: MFCD08062343
CAS-Nummer: 126120-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR