6-Methyl-4-(piperazin-1-yl)-2-(trifluoromethyl)quinoline 97%

Lieferant: Apollo Scientific
Gefahr

PC8526-1G PC8526-250MG
APOSPC8526-1GEA 0 EUR
APOSPC8526-1G APOSPC8526-250MG
6-Methyl-4-(piperazin-1-yl)-2-(trifluoromethyl)quinoline 97%
6-Methyl-4-(piperazin-1-yl)-2-(trifluoromethyl)quinoline
Formel: C₁₅H₁₆F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00728815
CAS-Nummer: 544429-25-6
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR