N-Methyl-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

PC8558-1G
APOSPC8558-1GEA 302 EUR
APOSPC8558-1G
N-Methyl-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenesulphonamide
N-Methyl-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenesulphonamide
Formel: C₁₃H₁₁F₃N₂O₃S
Molecular Weight: 332,3 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817448
CAS-Nummer: 952183-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR