2-[3-(Trifluoromethyl)phenyl]ethanethioamide
Lieferant: Apollo Scientific
|
Achtung
|
PC8559-1G
APOSPC8559-1GEA
234
EUR
APOSPC8559-1G
2-[3-(Trifluoromethyl)phenyl]ethanethioamide
2-[3-(Trifluoromethyl)phenyl]ethanethioamide
Formel:
C₉H₈F₃NS Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817423 CAS-Nummer: 952182-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...