4-Bromo-2-fluorophenylzinc iodide
Lieferant: Apollo Scientific
|
Achtung
|
PC8567-50ML
APOSPC8567-50MLEA
288
EUR
APOSPC8567-50ML
4-Bromo-2-fluorophenylzinc iodide
4-Bromo-2-fluorophenylzinc iodide
Formel:
C₆H₃BrFIZn Molecular Weight: 366,304 g/mol Siedepunkt: 65 °C Flammpunkt: 17,2 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02260131 CAS-Nummer: 352530-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...